EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Chemokines influence the migration and fate of neural precursor cells from the young adult and middle-aged rat subventricular zone.
PMID 22155482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB3418
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse