EMAIL THIS PAGE TO A FRIEND
Clinical nuclear medicine
Dicyclomine: cause of abnormally decreased gall bladder ejection with sincalide-stimulated hepatobiliary scintigraphy.
PMID 22157036
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7909
Dicyclomine hydrochloride, ≥99% (TLC), powder
C19H35NO2 · HCl