EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Effects of polychlorinated biphenyls in CD-1 mice: reproductive toxicity and intergenerational transmission.
PMID 22157103
Abstract
Several studies indicate that in utero and perinatal exposure to polychlorinated biphenyls (PCBs) induces adverse reproductive effects, but it remains unclear whether such effects may be transmitted to subsequent generations. We therefore investigated the association between maternal exposure to PCBs and reproductive health in male and female offspring over three generations. Mouse dams were fed 0, 1, 10, and 100 μg/kg/day of a PCB mixture (101 + 118) during pregnancy and lactation. PCB levels were measured in the tissues of both dams and offspring. PCB concentrations at all doses investigated were greater in the offspring than in the dams (p ≤ 0.0001) confirming that the progeny were exposed as a result of maternal exposure. In F1 offspring, exposure to PCBs resulted in reductions in (1) testis weight (p ≤ 0.05) and seminiferous tubule diameter (p ≤ 0.05), (2) sperm viability (p ≤ 0.0001) and developmental capacity (p ≤ 0.05), (3) ovary weight (p ≤ 0.05), (4) oocyte developmental capacity (p ≤ 0.05), and (5) increased follicular atresia (p ≤ 0.0001). In females, adverse effects were observed only in the F1 animals. In contrast, male offspring exhibited reduced sperm viability and altered seminiferous tubule distribution up to the third generation, showing intergenerational transmission. In summary, our data indicate that exposure to PCBs at the time of gonadal sex determination perturbed, significantly, the reproductive physiology of male and female offspring in adulthood. Furthermore, male reproductive deficiencies may be observed in at least two further generations. These findings have significant implications for reproductive health and fertility of animals and humans.
Related Materials