EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A bienzymatic amperometric immunosensor exploiting supramolecular construction for ultrasensitive protein detection.
PMID 22159310
Abstract
Self-assembly of a ferrocene-appended polymer bearing an antigen fragment and lactate oxidase on a cyclodextrin-modified surface provides a highly sensitive, easy-to-operate and self-sufficient immunosensor.