EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
First century of Chagas' disease: an overview on novel approaches to nifurtimox and benzonidazole delivery systems.
PMID 22161779
Abstract
Hundred years after the discovery of Chagas' disease, there is a lack of effective treatment to control this neglected disease caused by the parasite Trypanosoma cruzi. The transmission is primarily through vector-borne blood transfusion or during pregnancy, producing high mortality and morbidity among poor people in many countries of Latin America. In the last decades, the efforts have been focused mainly on the elimination of vectors. At the same time, screening of blood donors in order to avoid transfusional transmission has been improved all over the world. However, Chagas' disease is still a major public health problem, with estimates of nearly 90 million people at risk of infection and more than eight million infected in 18 endemic countries. Despite the high incidence in endemic regions and the dissemination of neglected diseases in North America and Europe, to date, there are only two drugs developed and prescribed for the treatment of Chagas' disease, nifurtimox (tablets of 120 mg) and benzonidazole (tablets of 100 mg). In this review, different approaches carried out in the last decades for developing novel pharmaceutical formulations for the delivery of nifurtimox and benznidazole are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3415
Nifurtimox, ≥98% (HPLC)
C10H13N3O5S