EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Secondary metabolite profile in induced tetraploids of wild Solanum commersonii Dun.
PMID 22162160
Abstract
The main aim of this work was to study the leaf secondary metabolite profiles of artificially induced tetraploids (2n=4x=48) of Solanum commersonii, a diploid (2n=2x=24) wild potato species. The tetraploid genotypes of S. commersonii were produced by oryzalin treatment. Both HPLC-UV and LC/MS analyses revealed that there were no qualitative differences in the metabolite profiles between the diploid S. commersonii and its tetraploids. By contrast, the results showed that the phenylpropanoid content was generally significantly higher in the tetraploids than in the diploid S. commersonii. Concerning the glycoalkaloids (GAs), the results provided evidence that the content of minor GAs (solanidenediol triose, solanidadienol lycotetraose, and solanidenol lycotetraose) was higher in tetraploids than in the diploid progenitor, while the content of major GAs (dehydrodemissine and dehydrocommersonine) was significantly higher in diploid S. commersonii than in its tetraploid genotypes. The results are discussed from the practical perspective of potato biodiversity enhancement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36182
Oryzalin, PESTANAL®, analytical standard
C12H18N4O6S
N12729
Oryzalin, analytical standard
C12H18N4O6S