EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
Enhanced bioavailability of sirolimus via preparation of solid dispersion nanoparticles using a supercritical antisolvent process.
PMID 22162657