EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica Hungarica
[Ontogeny of sulphonylurea-binding regulatory subunits of K(ATP) channels in the pregnant rat myometrium].
PMID 22165413
Abstract
K(ATP) channels are composed of sulphonylurea receptors (SURs) and potassium inward rectifiers (Kir(6.x)) that assemble to form a large octameric channel. This study was designed to examine the expression and role of sulphonylurea-binding regulatory subunits 1 [SUR1 (ABCC8)] and 2 [SUR2 (ABCC9)] of the K(ATP) channels in the pregnant rat myometrium with particular regard to the contractility. RT-PCR and Western blot analysis were performed to detect the presence of SUR1 and SUR2. The SUR1 levels were markedly increased in the early stages of pregnancy. The highest level was detected on day 6 of pregnancy, while in the late stages the levels of SUR1 were significantly decreased. The SUR2 level remained unchanged throughout pregnancy. The SUR-non-selective diazoxide and the SUR2-selective pinacidil inhibited oxytocin-induced contractions. Glibenclamide, a K(ATP) channel blocker, antagonized both pinacidil and diazoxide-induced relaxations. It was established that SURs are responsible for pharmacological reactivity of K(ATP) channel openers. We conclude that, both SURs are involved in the K(ATP) channel in the pregnant rat myometrium. It may further be concluded that "pinacidil-like" K(ATP) channel openers may be of therapeutic relevance as tocolytic agents in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P154
Pinacidil monohydrate, powder
C13H19N5 · H2O