EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Chirality sensing by a fluorescent binaphthocrown ether-polythiophene conjugate.
PMID 22167253
Abstract
Chiral recognition abilities of the title host for (R)- and (S)-α-methyl-4-nitrobenzylamine were examined in the ground and excited states to give a relative affinity (K(R)/K(S)) of 2.16 by spectral titration and a relative rate constant (k(R)/k(S)) of 2.23 by fluorescence quenching, revealing that the quenching process is static and not enantiodifferentiating.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
191434
4-Nitrobenzylamine hydrochloride, 97%
C7H8N2O2 · HCl