EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
CuCl/DABCO/4-HO-TEMPO-catalyzed aerobic oxidative synthesis of 2-substituted quinazolines and 4H-3,1-benzoxazines.
PMID 22168403
Abstract
The Cu/N-ligand/TEMPO catalytic system was first applied to the aerobic oxidative synthesis of heterocycles. As demonstrated, 2-substituted quinazolines and 4H-3,1-benzoxazines were synthesized efficiently from the one-pot reaction of aldehydes with 2-aminobenzylamines and 2-aminobenzyl alcohols, respectively, by employing CuCl/DABCO/4-HO-TEMPO as the catalysts and oxygen as the terminal oxidant.