EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Penta-O-galloyl-β-D-glucose attenuates cisplatin-induced nephrotoxicity via reactive oxygen species reduction in renal epithelial cells and enhances antitumor activity in Caki-2 renal cancer cells.
PMID 22172427
Abstract
Cisplatin shows limited therapeutic efficacy due to serious side effects such as nephrotoxicity and hepatotoxicity. In the present study, we demonstrate that 1,2,3,4,6-penta-O-galloyl-β-d-glucose (PGG) has protective effects against cisplatin-induced cytotoxicity and apoptosis in normal human primary renal epithelial cells (HRCs) while showing synergistic effect against cisplatin-induced cell death in human Caki-2 renal cancer cells. PGG significantly blocked cisplatin-mediated cytotoxicity and reduced cisplatin-induced sub-G1 accumulation in HRCs. Consistently, PGG reduced the number of apoptotic cell populations by TdT-mediated dUTP nick end labeling (TUNEL) and Live/Dead assays in cisplatin-treated HRCs. Furthermore, PGG suppressed PARP cleavage and caspase-3 activation, cytochrome c release, up-regulation of bax and p53 in cisplatin-treated HRCs. Moreover, PGG attenuated reactive oxygen species (ROS) production mediated by cisplatin treatment, suggesting that PGG prevented cisplatin-induced apoptosis by inhibiting ROS generation in HRCs. Notably, PGG significantly enhanced cytotoxicity and PARP cleavage in cisplatin-treated Caki-2 renal cancer cells. Combination Index (CI) revealed synergism between PGG and cisplatin in Caki-2 cells. Taken together, our findings suggest the dual effects of PGG as a protective supplement against cisplatin-induced toxicity in normal renal cells and a combination chemotherapeutic drug with cisplatin in renal cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7548
Penta-O-galloyl-β-D-glucose hydrate, ≥96% (HPLC)
C41H32O26 · xH2O