EMAIL THIS PAGE TO A FRIEND
Current opinion in microbiology
Histidine kinases and response regulators in networks.
PMID 22172627
Abstract
Two-component systems, composed of a histidine kinase (HK) and a response regulator (RR), are the major signal transduction devices in bacteria. Originally it was thought that these two components function as linear, phosphorylation-driven stimulus-response system. Here, we will review how accessory proteins are employed by HKs and RRs to mediate signal integration, scaffolding, interconnection and allosteric regulation, and how these two components are embedded in regulatory networks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53340
L-Histidine dihydrochloride, ≥99.0% (AT)
C6H9N3O2 · 2HCl