EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Preparation of three-bore hollow fiber charged nanofiltration membrane for separation of organics and salts.
PMID 22173422
Abstract
Three-bore hollow fiber charged nanofiltration (NF) membrane was prepared by interfacial polymerization (IP). The results showed that the flux and rejection of NF membrane prepared in this study increased with the increasing in the operating pressure. The water flux decreased and rejection for obvious dyes increased as the solute concentration increased. The separation factor for mixture of Xylenol orange/NaCl decreased when NaCl concentration in solution increased and could reach to as high as 18. In addition, three-bore hollow fiber charged nanofiltration membrane prepared in this study has excellent stability for strong acid (pH = 3), strong alkali (pH = 11) and high temperature solution (80 °C).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P23954
m-Phenylenediamine, flakes, 99%
C6H8N2
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
71360
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
CNa2O3 · 10H2O
13414
Sodium carbonate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · 10H2O
577782
Sodium carbonate decahydrate, 99.999% trace metals basis
CNa2O3 · 10H2O
S4132
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
CNa2O3 · H2O
13568
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · H2O
230952
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
CNa2O3 · H2O