EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of the pyridinyl analogues of dibenzylideneacetone (pyr-dba) via an improved Claisen-Schmidt condensation, displaying diverse biological activities as curcumin analogues.
PMID 22179573
Abstract
An efficient and easy procedure to synthesize the pyridinyl analogues of dibenzylideneacetone (pyr-dba) was developed by the condensation of substituted nicotinaldehyde and acetone in the presence of K(2)CO(3) in toluene-EtOH-H(2)O solvent system. Structurally diverse pyr-dba, including quinolinyl dba, can be prepared conveniently in moderate to excellent yields under mild conditions with this method. The resulting pyr-dba functioned as the enone analogs of curcumin and efficiently inhibited the activation of NF-κB and the growth of colorectal carcinoma HCT116 p53+/+ cells as well as the HIV-1 IN-LEDGF/p75 interaction.
Related Materials