EMAIL THIS PAGE TO A FRIEND
Bioresource Technology
Flocculation of Chlorella vulgaris induced by high pH: role of magnesium and calcium and practical implications.
PMID 22182473
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223484
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
CNa2O3
222321
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
CNa2O3
223530
Sodium carbonate, powder, ≥99.5%, ACS reagent
CNa2O3
S2127
Sodium carbonate, ReagentPlus®, ≥99.5%
CNa2O3
31432
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
CNa2O3
13418
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
CNa2O3
900503
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
900505
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
CNa2O3
900504
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
CNa2O3
71360
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
CNa2O3 · 10H2O
13414
Sodium carbonate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · 10H2O
577782
Sodium carbonate decahydrate, 99.999% trace metals basis
CNa2O3 · 10H2O
S4132
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
CNa2O3 · H2O
13568
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
CNa2O3 · H2O
230952
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
CNa2O3 · H2O