EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Effect of chronic administration of estradiol, progesterone, and tibolone on the expression and phosphorylation of glycogen synthase kinase-3β and the microtubule-associated protein tau in the hippocampus and cerebellum of female rat.
PMID 22183707
Abstract
Gonadal hormones regulate expression and activation of protein tau. Tibolone is a drug used as first- choice comprehensive treatment for the relief of menopausal symptoms, because it and its various metabolites have estrogenic properties and progestogenic/androgenic effects; however, the effect on the activation of tau protein and its signaling cascade in the brain is unknown. We studied the effect of chronic administration of estradiol (E2), progesterone (P4), and tibolone (TIB) on the expression and phosphorylation of microtubule-associated protein tau and glycogen synthase kinase-3β (GSK3β) in the hippocampus and cerebellum of ovariectomized rats. Ovariectomized adult female rats were implanted with pellets of vehicle, E2, or P4 or were treated with TIB by oral administration for 60 days. The animals were sacrificed, and tissue proteins were analyzed by Western blot. We observed that, in the hippocampus, administration of E2, P4, or TIB significantly decreased the protein content of hyperphosphorylated tau and increased the tau dephosphorylated form, whereas only treatment with TIB increased the content of the phosphorylated form of GSK3β. In the cerebellum, E2 and TIB treatments resulted in a significant decrease in the expression of hyperphosphorylated tau, whereas E2 and TIB increased phosphorylated GSK3β; P4 had no effect. These results indicate that chronic administration of gonadal hormones and tibolone modulates tau and GSK3β phosphorylation in hippocampus and cerebellum of the rat and may exert a neuroprotective effect in these tissues.
Related Materials