EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Polychlorinated biphenyl congeners that increase the glucuronidation and biliary excretion of thyroxine are distinct from the congeners that enhance the serum disappearance of thyroxine.
PMID 22187485
Abstract
Polychlorinated biphenyl (PCB) congeners differentially reduce serum thyroxine (T(4)) in rats, but little is known about their ability to affect biliary excretion of T(4). Thus, male Sprague-Dawley rats were orally administered Aroclor-1254, Aroclor-1242 (32 mg/kg per day), PCB-95, PCB-99, PCB-118 (16 mg/kg per day), PCB-126 (40 μg/kg per day), 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) (3.9 μg/kg per day), or corn oil for 7 days. Twenty-four hours after the last dose, [(125)I]T(4) was administered intravenously, and blood, bile, and urine samples were collected for quantifying [(125)I]T(4) and in bile [(125)I]T(4) metabolites. Serum T(4) concentrations were reduced by all treatments, but dramatic reductions occurred in response to Aroclor-1254, PCB-99 [phenobarbital (PB)-type congener], and PCB-118 (mixed-type congener). None of the treatments increased urinary excretion of [(125)I]T(4). Aroclor-1254, PCB-118, TCDD, and PCB-126 (TCDD-type congener) increased biliary excretion of T(4)-glucuronide by 850, 756, 710, and 573%, respectively, corresponding to marked induction of hepatic UDP-glucuronosyltransferase (UGT) activity toward T(4). PCB-95 and PCB-99 did not induce UGT activity; therefore, the increased biliary excretion of T(4)-glucuronide was related to the affinity of congeners for the aryl hydrocarbon receptor. The disappearance of [(125)I]T(4) from serum was rapid (within 15-min) and was increased by Aroclor-1254, PCB-99 and PCB-118. Thus, reductions in serum T(4) in response to PCBs did not always correspond with UGT activity toward T(4) or with increased biliary excretion of T(4)-glucuronide. The rapid disappearance of [(125)I]T(4) from the serum of rats treated with PB-like PCBs suggests that increased tissue uptake of T(4) is an additional mechanism by which PCBs may reduce serum T(4).