EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Effects of the selective 5-HT(7) receptor antagonist SB-269970 in animal models of psychosis and cognition.
PMID 22189656
Abstract
The 5-hydroxytryptamine7 (5-HT7) receptor is a G-protein coupled receptor for serotonin that has been implicated in the pathophysiology of psychiatric and neurological disorders including anxiety, depression and schizophrenia. A number of studies have attempted to evaluate the potential role of the 5-HT7 receptor in schizophrenia by utilising genetic or pharmacological tools but to date these have provided conflicting results. Here we investigate the effect of a selective 5-HT7 receptor antagonist, SB-269970, in in vivo psychosis and cognition models and relate efficacy to brain exposures of the compound. SB-269970 significantly attenuated amphetamine-induced rearing and circling in rats. A similar effect was observed in an N-methyl d-aspartic acid (NMDA) receptor antagonist driven psychosis model, where SB-269970 significantly reversed phencyclidine-induced hyperlocomotion, rearing and circling; although the effect was not as robust as with the 5-HT2a receptor antagonist positive control, MDL100,907. SB-269970 also attenuated a temporal deficit in novel object recognition (NOR), indicative of an improvement in recognition memory. Pharmacokinetic analysis of plasma and brain samples taken after behavioural testing confirmed that efficacy was achieved at doses and pre-treatment times where receptor occupancy was substantial. These findings highlight the anti-psychotic and pro-cognitive potential of 5-HT7 receptor antagonists and warrant further studies to explore their therapeutic potential in schizophrenia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3324
M100907, ≥98% (HPLC)
C22H28FNO3