EMAIL THIS PAGE TO A FRIEND
Revista brasileira de psiquiatria (Sao Paulo, Brazil : 1999)
Use of anticholinergic drugs and worsening of pemphigus foliaceus in a patient with bipolar disorder.
PMID 22189933
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5311
Biperiden hydrochloride, ≥98% (HPLC), powder
C21H30ClNO