EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Mercapturic acid formation in cultured opossum kidney cells.
PMID 2219181
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B19800
S-Benzyl-L-cysteine, 97%
C10H13NO2S