EMAIL THIS PAGE TO A FRIEND
Phytochemistry
meta-Tyrosine in Festuca rubra ssp. commutata (Chewings fescue) is synthesized by hydroxylation of phenylalanine.
PMID 22192329
Abstract
m-Tyrosine is a non-protein amino acid that is structurally similar to the common protein amino acids p-tyrosine and phenylalanine. Copious amounts of m-tyrosine can be found in root exudates of the fine fescue cultivar, Festuca rubra L. ssp. commutata (Chewings fescue). The phytotoxicity of m-tyrosine may contribute to the allelopathic potential of F. rubra. m-Tyrosine in Euphorbia myrsinites (donkey-tail spurge), was previously shown to be synthesized via transamination of m-hydroxyphenylpyruvate. Here we show that m-tyrosine biosynthesis in F. rubra occurs through direct hydroxylation of phenylalanine in the root tips, perhaps through the activity of a cytochrome P450 enzyme. Hence, E. myrsinites and F. rubra, the only two plant species known to produce m-tyrosine, use distinct biosynthetic pathways that likely arose independently in evolutionary history.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3629
DL-m-Tyrosine, crystalline
C9H11NO3