EMAIL THIS PAGE TO A FRIEND
FEBS letters
cGMP and cyclic nucleotide-gated channels participate in mouse sperm capacitation.
PMID 22192355
Abstract
During capacitation of mammalian sperm intracellular [Ca(2+)] and cyclic nucleotides increase, suggesting that CNG channels play a role in the physiology of sperm. Here we study the effect of capacitation, 8Br-cAMP (8-bromoadenosine 3',5'-cyclic monophosphate) and 8Br-cGMP (8-bromoguanosine 3',5'-cyclic monophosphate) on the macroscopic ionic currents of mouse sperm, finding the existence of different populations of sperm, in terms of the recorded current and its response to cyclic nucleotides. Our results show that capacitation and cyclic nucleotides increase the ionic current, having a differential sensitivity to cGMP (cyclic guanosine monophosphate) and cAMP (cyclic adenosine monophosphate). Using a specific inhibitor we determine the contribution of CNG channels to macroscopic current and capacitation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1081
8-Chloroadenosine 3′,5′-cyclic-monophosphate, ≥85%
C10H11ClN5O6P