EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Influence of halogen substituents on the catalytic oxidation of 2,4,6-halogenated phenols by Fe(III)-tetrakis(p-hydroxyphenyl) porphyrins and potassium monopersulfate.
PMID 22193213
Abstract
The influence of halogen substituents on the catalytic oxidation of 2,4,6-trihalogenated phenols (TrXPs) by iron(III)-porphyrin/KHSO₅ catalytic systems was investigated. Iron(III)-5,10,15,20-tetrakis(p-hydroxyphenyl)porphyrin (FeTHP) and its supported variants were employed, where the supported catalysts were synthesized by introducing FeTHP into hydroquinone-derived humic acids via formaldehyde poly-condensation. F (TrFP), Cl (TrCP), Br (TrBP) and I (TrIP) were examined as halogen substituents for TrXPs. Although the supported catalysts significantly enhanced the degradation and dehalogenation of TrFP and TrCP, the oxidation of TrBP and TrIP was not enhanced, compared to the FeTHP catalytic system. These results indicate that the degree of oxidation of TrXPs is strongly dependent on the types of halogen substituent. The order of dehalogenation levels for halogen substituents in TrXPs was F > Cl > Br > I, consistent with their order of electronegativity. The electronegativity of a halogen substituent affects the nucleophilicity of the carbon to which it is attached. The levels of oxidation products in the reaction mixtures were analyzed by GC/MS after extraction with n-hexane. The most abundant dimer product from TrFP via 2,6-difluoroquinone is consistent with a scenario where TrXP, with a more electronegative halogen substituent, is readily oxidized, while less electronegative halogen substituents are oxidized less readily by iron(III)-porphyrin/KHSO₅ catalytic systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2