EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Phonon dynamics and electron-phonon coupling in pristine picene.
PMID 22193510
Abstract
The paper reports a complete analysis of the phonon structure of crystalline picene, a recently announced organic semiconductor. Both lattice and intramolecular vibrations are investigated. An exhaustive assignment of lattice phonons is obtained through polarized Raman spectra assisted by lattice dynamics calculations based on a well tested atom-atom potential model. Raman, infrared spectra and density functional (DFT) calculations are used for the characterization of intramolecular modes. Coupling between low-frequency molecular vibrations and lattice phonons is accounted for. Molecule-to-molecule transfer integrals, as well as the Peierls and Holstein (non-local and local) coupling constants, are evaluated through the semiempirical method INDO/S (Intermediate Neglect of Differential Overlap with Spectroscopic parametrization).