EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Highly selective recognition on enatioselective of R(+) and S(-)-1,1-Bi(2-naphthol) using of Eu(dap)3 complex as photo probe.
PMID 22194002
Abstract
The chiral recognition phenomenon was observed in enantioselective and excited-state energy transfer processes. Based on bimolecular luminescence quenching kinetics for a system containing chiral molecules, the quenching efficiency was evaluated by Stern-Volmer equation for a system containing a chiral R(+) and S(-) resolved quencher species. The utility of this methodology is confirmed by examining the enantio-selective excited-state quenching between Eu(dpa)(3) complex (where dpa = pyridine-2,6-dicarboxylate) acting as the energy donor to R(+)-1,1-Bi(2-Naphthol) and S(-)-1,1-Bi(2-naphthol) as the energy acceptor was studied in solution. The results of this study confirm the utility of luminescence measurements as a probe of chiral discriminatory behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104655
1,1′-Bi-2-naphthol, 99%
C20H14O2