PloS one
EZH2 codon 641 mutations are common in BCL2-rearranged germinal center B cell lymphomas.
PMID 22194861
Abstract
Mutations at codon 641 of EZH2 are recurrent in germinal center B cell lymphomas, and the most common variants lead to altered EZH2 enzymatic activity and enhanced tri-methylation of histone H3 at lysine 27, a repressive chromatin modification. As an initial step toward screening patients for cancer genotype-directed therapy, we developed a screening assay for EZH2 codon 641 mutations amenable for testing formalin-fixed clinical specimens, based on the sensitive SNaPshot single nucleotide extension technology. We detected EZH2 mutations in 12/55 (22%) follicular lymphomas (FL), 5/35 (14%) diffuse large B cell lymphomas with a germinal center immunophenotype (GCB-DLBCL), and 2/11 (18%) high grade B cell lymphomas with concurrent rearrangements of BCL2 and MYC. No EZH2 mutations were detected in cases of Burkitt lymphoma (0/23). EZH2 mutations were frequently associated with the presence of BCL2 rearrangement (BCL2-R) in both the FL (28% of BCL-R cases versus 0% of BCL2-WT cases, p<0.05) and GCB-DLBCL groups (33% of BCL2-R cases versus 4% of BCL2-WT cases, p<0.04), and across all lymphoma types excluding BL (27% of BCL2-R cases versus 3% of BCL2-WT cases, p<0.003). We confirmed gain-of-function activity for all previously reported EZH2 codon 641 mutation variants. Our findings suggest that EZH2 mutations constitute an additional genetic "hit" in many BCL2-rearranged germinal center B cell lymphomas. Our work may be helpful in the selection of lymphoma patients for future trials of pharmacologic agents targeting EZH2 and EZH2-regulated pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody