EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Discovering small molecules that promote cardiomyocyte generation by modulating Wnt signaling.
PMID 22195568
Abstract
We have developed a robust inxa0vivo small-molecule screen that modulates heart size and cardiomyocyte generation in zebrafish. Three structurally related compounds (Cardionogen-1 to Cardionogen-3) identified from our screen enlarge the size of the developing heart via myocardial hyperplasia. Increased cardiomyocyte number in Cardionogen-treated embryos is due to expansion of cardiac progenitor cells.xa0In zebrafish embryos and murine embryonic stem (ES) cells, Cardionogen treatment promotes cardiogenesis during and after gastrulation, whereas it inhibits heart formation before gastrulation. Cardionogen-induced effects can be antagonized by increasing Wnt/β-catenin signaling activity. Wexa0demonstrate that Cardionogen inhibits Wnt/β-catenin-dependent transcription in murine ES cells and zebrafish embryos. Cardionogen can rescue Wnt8-induced cardiomyocyte deficiency and heart-specific phenotypes during development. These findings demonstrate that inxa0vivo small-molecule screens targeting heart size can reveal compounds with cardiomyogenic effects and identify underlying target pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0458
Cardionogen-1, ≥98% (HPLC)
C13H14N4OS
SML0427
Cardionogen-2, ≥98% (HPLC)
C16H13N5O2S