EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Drug carriers based on cyclodextrin inclusion complexes for the controlled release of hydrophobic drugs.
PMID 22195793
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399396
Benzoin, purified by sublimation, ≥99.5%
C14H12O2
B8681
Benzoin, 98%
C14H12O2