EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Self-assembly of chitosan-g-PEG and α-cyclodextrin into hollow spheres in aqueous solution.
PMID 22195855