EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Preliminary study of the relationship between water absorbency and zeta potentials of crosslinked poly(acrylic acid).
PMID 22195899
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2