EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Novel pH-sensitive zwitterionic poly(amino acid) derivatives for drug delivery.
PMID 22195952