EMAIL THIS PAGE TO A FRIEND
Biological psychiatry
The stress-induced cytokine interleukin-6 decreases the inhibition/excitation ratio in the rat temporal cortex via trans-signaling.
PMID 22196984
Abstract
Although it is known that stress elevates the levels of pro-inflammatory cytokines and promotes hyper-excitable central conditions, a causal relationship between these two factors has not yet been identified. Recent studies suggest that increases in interleukin 6 (IL-6) levels are specifically associated with stress. We hypothesized that IL-6 acutely and directly induces cortical hyper-excitability by altering the balance between synaptic excitation and inhibition. We used patch-clamp to determine the effects of exogenous or endogenous IL-6 on electrically evoked postsynaptic currents on a cortical rat slice preparation. We used control subjects or animals systemically injected with lipopolysaccharide or subjected to electrical foot-shock as rat models of stress. In control animals, IL-6 did not affect excitatory postsynaptic currents but selectively and reversibly reduced the amplitude of inhibitory postsynaptic currents with a postsynaptic effect. The IL-6-induced inhibitory postsynaptic currents decrease was inhibited by drugs interfering with receptor trafficking and/or internalization, including wortmannin, Brefeldin A, 2-Br-hexadecanoic acid, or dynamin peptide inhibitor. In both animal models, stress-induced decrease in synaptic inhibition/excitation ratio was prevented by prior intra-ventricular injection of an analog of the endogenous IL-6 trans-signaling blocker gp130. Our results suggest that stress-induced IL-6 shifts the balance between synaptic inhibition and excitation in favor of the latter, possibly by decreasing the density of functional γ-aminobutyric acid A receptors, accelerating their removal and/or decreasing their insertion rate from/to the plasma membrane. We speculate that this mechanism could contribute to stress-induced detrimental long-term increases in central excitability present in a variety of neurological and psychiatric conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21604
2-Bromohexadecanoic acid, ≥99.0% (GC)
C16H31BrO2
238422
2-Bromohexadecanoic acid, ~97%
C16H31BrO2