EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Collagenase-1 injection improved tumor distribution and gene expression of cationic lipoplex.
PMID 22197775
Abstract
Elevated interstitial fluid pressure (IFP) in a tumor is a barrier to tumor accumulation of systemic delivery of nanocarriers. In this study, we investigated whether intravenous injection of type I collagenase (collagenase-1) reduced IFP in tumors and increased the accumulation and gene expression of cationic liposome/plasmid DNA complex (lipoplex) in tumors after intravenous injection into mice bearing mouse lung carcinoma LLC tumors. Collagenase-1 reduced the amount of type I collagen in the tumor, and significantly decreased IFP by 65% at 1h after injection. Therefore, collagenase-1 induced 1.5-fold higher accumulation and 2-fold higher gene expression of lipoplex in tumors after intravenous injection. These findings indicated that intravenous injection of collagenase-1 improved the accumulation of lipoplex by decreasing IFP in tumors. These results support the potential use of collagen digestion as a strategy to improve systemic gene delivery into tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1889
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C0773
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
C1639
Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C1764
Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid
C9722
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
C5894
Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
C9697
Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
C7926
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
C2399
Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
C8051
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
C0255
Collagenase from Clostridium histolyticum, purified by chromatography, ≥0.25 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
C2014
Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
C2799
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
C8176
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid