EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section E, Structure reports online
N-Benzyl-P-(2-ethyl-phen-yl)-P-phenyl-phosphinic amide.
PMID 22199881
Abstract
In the crystal structure of the title compound, C(21)H(22)NOP, the amine H atom is involved in N-H⋯O hydrogen-bonding inter-actions, resulting in chains along the c axis. The crystal lattice is consolidated by weak inter-molecular C-H⋯π inter-actions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343315
1-Bromo-2-ethylbenzene, 99%
C8H9Br