EMAIL THIS PAGE TO A FRIEND
BMC research notes
A refined, rapid and reproducible high resolution melt (HRM)-based method suitable for quantification of global LINE-1 repetitive element methylation.
PMID 22204640
Abstract
The methylation of DNA is recognized as a key mechanism in the regulation of genomic stability and evidence for its role in the development of cancer is accumulating. LINE-1 methylation status represents a surrogate measure of genome-wide methylation. Using high resolution melt (HRM) curve analysis technology, we have established an in-tube assay that is linear (r > 0.9986) with a high amplification efficiency (90-105%), capable of discriminating between partcipant samples with small differences in methylation, and suitable for quantifying a wide range of LINE-1 methylation levels (0-100%)--including the biologically relevant range of 50-90% expected in human DNA. We have optimized this procedure to perform using 2 μg of starting DNA and 2 ng of bisulfite-converted DNA for each PCR reaction. Intra- and inter-assay coefficients of variation were 1.44% and 0.49%, respectively, supporting the high reproducibility and precision of this approach. In summary, this is a completely linear, quantitative HRM PCR method developed for the measurement of LINE-1 methylation. This cost-efficient, refined and reproducible assay can be performed using minimal amounts of starting DNA. These features make our assay suitable for high throughput analysis of multiple samples from large population-based studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15236
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)
C7H19NO2Si2