EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Practical synthesis of hydroxychromenes and evaluation of their biological activity.
PMID 22205090
Abstract
A simple and efficient seven steps synthesis of brodifacoum and difethialone from phenylacetyl chloride is described. The key synthetic strategies involve Friedel-Crafts acylation, intramolecular ring cyclization and condensation reaction in the presence of Brønsted-Lowry acids. It was found that brodifacoum showed favorable inhibiting activities on LPS-stimulated nitrite production in BV-2 microglia cells. Brodifacoum exhibited superior anti-inflammatory effects than difethialone. We expect that an efficient linear synthesis of 4-hydroxycoumarin derivatives and key fragments that are useful agents for the modulation of inflammation as well as inhibition of coagulation will be of practical use.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46036
Brodifacoum, PESTANAL®, analytical standard
C31H23BrO3