EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Suspected unexpected adverse effect of sugammadex: hypotension.
PMID 22205274
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2451000 Etilefrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C10H15NO2 · HCl