EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Development of iodoacetic acid-based cysteine mass tags: detection enhancement for cysteine-containing peptide by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
PMID 22206935
Abstract
We developed and characterized 6 new cysteine mass tags for high-sensitivity peptide analysis. The structural features are: (1) iodoacetyl group for thiol tagging, (2) hydrophilic character for reducing sample loss, (3) tertiary amino, quaternary ammonium, or guanidino group for high proton affinity, and (4) no amide bonding for minimizing fragmentation of tag moiety in collision-induced dissociation. By using these tags, 2- to 200-fold MS sensitivity was achieved, compared to control peptide with carbamydomethylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I4386
Iodoacetic acid, ≥98.0% (T)
C2H3IO2