EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereoselective α-quaternization of 3-methoxycycloalk-2-enones via 1,4-diastereoinduction of alkoxy dienolates.
PMID 22208771
Abstract
The alkylation of dienolates generated from 3-methoxycycloalk-2-enones having a 3'-hydroxyl alkenyl chain provides the corresponding quaternized cycloalkenones in a highly diastereoselective manner. The high degree of stereocontrol in the α-quaternization possibly implies intervention of a rigid chelating transition state that allows an efficient 1,4-asymmetric induction to take place.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
393274
3-Methoxy-2-cyclopenten-1-one, 99%
C6H8O2