EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Production of indole-3-acetic acid via the indole-3-acetamide pathway in the plant-beneficial bacterium Pseudomonas chlororaphis O6 is inhibited by ZnO nanoparticles but enhanced by CuO nanoparticles.
PMID 22210218
Abstract
The beneficial bacterium Pseudomonas chlororaphis O6 produces indole-3-acetic acid (IAA), a plant growth regulator. However, the pathway involved in IAA production in this bacterium has not been reported. In this paper we describe the involvement of the indole-3-acetamide (IAM) pathway in IAA production in P. chlororaphis O6 and the effects of CuO and ZnO nanoparticles (NPs). Sublethal levels of CuO and ZnO NPs differentially affected the levels of IAA secreted in medium containing tryptophan as the precursor. After 15 h of growth, CuO NP-exposed cells had metabolized more tryptophan than the control and ZnO NP-challenged cells. The CuO NP-treated cells produced higher IAA levels than control cultures lacking NPs. In contrast, ZnO NPs inhibited IAA production. Mixing of CuO and ZnO NPs resulted in an intermediate level of IAA production relative to the levels in the separate CuO and ZnO NP treatments. The effect of CuO NPs on IAA levels could be duplicated by ions at the concentrations released from the NPs. However, ion release did not account for the inhibition caused by the ZnO NPs. The mechanism underlying changes in IAA levels cannot be accounted for by effects on transcript accumulation from genes encoding a tryptophan permease or the IAM hydrolase in 15-h cultures. These findings raise the issue of whether sublethal doses of NPs would modify the beneficial effects of association between plants and bacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286281
Indole-3-acetamide, 98%
C10H10N2O