EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
Activation of the unfolded protein response pathway causes ceramide accumulation in yeast and INS-1E insulinoma cells.
PMID 22210926
Abstract
Sphingolipids are not only important components of membranes but also have functions in protein trafficking and intracellular signaling. The LCB1 gene encodes a subunit of the serine palmitoyltransferase, which is responsible for the first step of sphingolipid synthesis. Here, we show that activation of the unfolded protein response (UPR) can restore normal ceramide levels and viability in yeast cells with a conditional defect in LCB1. Dependence on UPR was demonstrated by showing the HAC1-dependence of the suppression. A similar induction of ceramides by UPR seems to take place in mammalian cells. In rat pancreatic INS-1E cells, UPR activation induces the transcription of the CerS6 gene, which encodes a ceramide synthase. This correlates with the specific accumulation of ceramide with a C16 fatty acyl chain upon UPR activation. Therefore, our study reveals a novel connection between UPR induction and ceramide synthesis that seems to be conserved between yeast and mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43799
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)
C34H67NO3