EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
TcyR regulates L-cystine uptake via the TcyABC transporter in Streptococcus mutans.
PMID 22212096
Abstract
Streptococcus mutans, a primary dental pathogen, has a remarkable capacity to scavenge nutrients from the oral biofilm for its survival. Cystine is an amino acid dimer formed by the oxidation of two cysteine residues that is required for optimal growth of S. mutans, which modulates l-cystine uptake via two recently identified transporters designated TcyABC and TcyDEFGH, which have not been fully characterized. Using a nonpolar tcyABC-deficient mutant (SmTcyABC), here, we report that l-cystine uptake is drastically diminished in the mutant, whereas its ability to grow is severely impaired under l-cystine starvation conditions, relative to wild type. A substrate competition assay showed that l-cystine uptake by the TcyABC transporter was strongly inhibited by dl-cystathionine and l-djenkolic acid and moderately inhibited by S-methyl-l-cysteine and l-cysteine. Using gene expression analysis, we observed that the tcyABC operon was upregulated under cystine starvation. TcyABC has been shown to be positively regulated by the LysR-type transcriptional regulator CysR. We identified another LysR-type transcriptional regulator that negatively regulates TcyABC with homology to the Bacillus subtilis YtlI regulator, which we termed TcyR. Our study enhances the understanding of l-cystine uptake in S. mutans, which allows survival and persistence of this pathogen in the oral biofilm.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7505
L-Cystathionine, ≥98% (TLC)
C7H14N2O4S