EMAIL THIS PAGE TO A FRIEND
Biochemistry
Structure of the apo form of Bacillus stearothermophilus phosphofructokinase.
PMID 22212099
Abstract
The crystal structure of the unliganded form of Bacillus stearothermophilus phosphofructokinase (BsPFK) was determined using molecular replacement to 2.8 Å resolution (Protein Data Bank entry 3U39 ). The apo BsPFK structure serves as the basis for the interpretation of any structural changes seen in the binary or ternary complexes. When the apo BsPFK structure is compared with the previously published liganded structures of BsPFK, the structural impact that the binding of the ligands produces is revealed. This comparison shows that the apo form of BsPFK resembles the substrate-bound form of BsPFK, a finding that differs from previous predictions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3627
D-Fructose 6-phosphate disodium salt hydrate, ≥98%, amorphous powder
C6H11Na2O9P · xH2O