EMAIL THIS PAGE TO A FRIEND
Experimental gerontology
Reserpine modulates neurotransmitter release to extend lifespan and alleviate age-dependent Aβ proteotoxicity in Caenorhabditis elegans.
PMID 22212533
Abstract
Aging is a debilitating process often associated with chronic diseases such as diabetes, cardiovascular and neurodegenerative diseases like Alzheimer's disease (AD). AD occurs at a very high incidence posing a huge burden to the society. Model organisms such as C. elegans become essential to understand aging or lifespan extension - the etiology, molecular mechanism and identification of new drugs against age associated diseases. The AD model, manifesting Aβ proteotoxicity, in C. elegans is well established and has provided valuable insights. Earlier, we have reported that Reserpine, an FDA-approved antihypertensive drug, increases C. elegans lifespan with a high quality of life and ameliorates Aβ toxicity in C. elegans. But reserpine does not seem to act through the known lifespan extension pathways or inhibition of its known target, vesicular monoamine transporter, VMAT. Reserpine's mode of action and the pathways it activates are not known. Here, we have evaluated the presynaptic neurotransmitter(s) release pathway and identified acetylcholine (ACh) as the crucial player for reserpine's action. The corroborating evidences are: i) lack of lifespan extension in the ACh loss of function (hypomorphic) - synthesis (cha-1) and transport (unc-17) mutants; ii) mitigation of chronic aldicarb effect; iii) lifespan extension in dopamine (cat-2) and dopamine and serotonin (bas-1) biosynthetic mutants; iv) no rescue from exogenous serotonin induced paralysis in the AD model worms; upon reserpine treatment. Thus, modulation of acetylcholine is essential for reserpine's action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33386
Aldicarb, PESTANAL®, analytical standard
C7H14N2O2S