EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Tuning fluorescent response of nanoscale film with polymer grafting.
PMID 22213257
Abstract
An effective method for tuning fluorescent response of an ultrathin (5 nm) polymer film, which can be used for generation of sensing arrays, is reported. This method is distinctive in that the modification of the optical response is achieved with polymer grafting of a non-fluorescent polymer to a fluorescent film. Using this approach, a number of films demonstrating different fluorescent emission when exposed to solvent vapors were synthesized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
705314
Poly(glycidyl methacrylate), Mn 10,000-20,000