EMAIL THIS PAGE TO A FRIEND
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases
[Clinical analysis of acute bromadiolone poisoning].
PMID 22214168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46035
Bromadiolon, PESTANAL®, analytical standard
C30H23BrO4