EMAIL THIS PAGE TO A FRIEND
International ophthalmology
Bilateral methicillin-resistant Staphylococcus aureus keratitis following hyperopic photorefractive surgery.
PMID 22215418
Abstract
To report clinical manifestations of a female patient with bilateral bacterial keratitis following photorefractive keratectomy (PRK). Bilateral PRK was performed for moderate hyperopia. Bandage contact lenses were fitted at the conclusion of the surgery. Bilateral infectious keratitis with hypopion was diagnosed within 4 days after surgery. Smear and culture were obtained and showed the presence of methicillin-resistant Staphylococcus aureus (MRSA). The patient was treated with systemic prednisone and topical antibiotics (vancomycin, tobramycin and netylmicin) and betamethasone. After 1 month corneal leukoma was still present and remained unchanged during the following 7 months. Infectious keratitis is a rare complication of PRK that appears early in the postoperative period. MRSA keratitis may determine long-term visual impairment despite prompt therapeutic intervention.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0755 Netilmicin sulfate salt
C21H41N5O7 · 2.5H2O4S