EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly stereoselective synthesis of cis-β-enaminones mediated by diethyl azodicarboxylate.
PMID 22218457
Abstract
Promoted by diethyl azodicarboxylate, a novel and highly stereoselective synthesis of cis-β-enaminones via oxidative dehydrogenation and hydration of the substituted propargylamines was realized. The possible mechanism was also proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50900
Propargylamine, 98%
C3H5N
P50919
Propargylamine hydrochloride, 95%
C3H5N · HCl