EMAIL THIS PAGE TO A FRIEND
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery
CD26/DPP-4 inhibition recruits regenerative stem cells via stromal cell-derived factor-1 and beneficially influences ischaemia-reperfusion injury in mouse lung transplantation.
PMID 22219460
Abstract
The CD26 antigen is a transmembrane glycoprotein that is constitutively expressed on activated lymphocytes and in pulmonary parenchyma. This molecule is also identified as dipeptidyl peptidase-4 (DPP-4) that cleaves a host of biologically active peptides. Here, we aimed to identify an important substrate of CD26/DPP-4-stromal cell-derived factor-1 (SDF-1/CXCL12)-as a key modulator for stem-cell homing together with its receptor CXCR4 in response to ischaemic injury of the lung. Orthotopic single lung transplantation (Tx) was performed between syngeneic C57BL/6 mice. Inhibition of CD26/DPP-4 activity in recipients was achieved using vildagliptin (10 mg/kg, every 12 h) subcutaneously, and 6 h ischaemia time was applied prior to implantation. Forty-eight hours after Tx, lung histology, SDF-1 levels (enzyme-linked immunosorbent assay) in lung, spleen and plasma, and expression of the SDF-1 receptor CXCR4 in blood and lung were assessed. Homing of regenerative progenitor cells to the transplanted lung was evaluated using fluorescent-activated cell sorting. Compared with untreated lung transplanted mice, systemic DPP-4 inhibition of Tx recipients resulted in an increase in protein concentration of SDF-1 in plasma, spleen and lung. Concordantly, the frequency of cells bearing the SDF-1 receptor CXCR4 rose significantly in the circulation and also in the lungs of DPP-4-inhibited recipients. We found co-expression of CXCR4/CD34 in the grafts of animals treated with vildagliptin, and the stem-cell markers Flt-3 and c-kit were present on a significantly increased number of cells. The morphology of grafts from DPP-4 inhibitor-treated recipients revealed less alveolar oedema when compared with untreated recipients. Targeting the SDF-1-CXCR4 axis through CD26/DPP-4 inhibition increased the intragraft number of progenitor cells contributing to the recovery from ischaemia-reperfusion lung injury. Stabilization of endogenous SDF-1 is achievable and may be a promising strategy to intensify sequestration of regenerative stem cells and thus emerges as a novel therapeutic concept.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2714 Protease Inhibitor Cocktail powder, for general use, lyophilized powder