EMAIL THIS PAGE TO A FRIEND
Archives of gynecology and obstetrics
Pancytopenia in a patient given pyrimethamine and sulphamethoxidiazine during pregnancy.
PMID 2221996
Abstract
This is a report of a pregnant woman who became pancytopenic in the second trimester following pyrimethamine administration. The patient recovered on steroid, antibiotics, transfusion, folic acid and folinic acid therapy, and the pregnancy was uneventful afterwards. The patient was delivered of a healthy term-infant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46834
Sulfameter, VETRANAL®, analytical standard
C11H12N4O3S
S0383
Sulfameter
C11H12N4O3S